Learn More
Thermo Scientific™ Labtainer™ 2D BioProcess Container (BPC), Aegis™ 5-14 film, three ports, Luer locks
GREENER_CHOICE
Código de producto. 17860667
Change view
Click to view available options
Capacidad (métrico):
50 mL
100 mL
250 mL
500 mL
1 L
2000 mL
2 L
5 L
10 L
20 L
50 L
Tamaño de la unidad:
paquete de 10
Código de producto. Capacidad (métrico) unitSize
17860667 50 L paquete de 10
17870607 50 mL paquete de 10
17880667 100 mL paquete de 10
17800677 250 mL paquete de 10
17890607 500 mL paquete de 10
17800617 1 L paquete de 10
17820677 2000 mL paquete de 10
17840607 5 L paquete de 10
17870667 10 L paquete de 10
17880607 20 L paquete de 10
17890667 2 L paquete de 10
11 options
Código de producto. 17860667
Marca: Thermo Scientific™ SH31049.04
Thermo Scientific Labtainer 2D BioProcess Containers (BPCs) are composed of Aegis 5-14 single-web, multi-layer film.
Features of the Labtainer 2D BPC include:
• Helps eliminate post-use cleaning steps required with re-useable containers and reduce cross-contamination risks
• Constructed in an ISO 7 cleanroom under cGMP conditions
Applications of the Labtainer 2D BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors
• Prepare and store buffer, culture media, and process liquid
• Collect chromatography feed and fractions
• Harvest, storage, and transport bulk drug product and bulk drug precursors
Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands
TRUSTED_SUSTAINABILITY
Especificaciones
Esterilizables en autoclave Not Autoclavable
Capacidad (métrico) 50 L
Certificaciones/conformidad ISO 7
Color Transparente
Desechables Single-use
Para utilizar con (aplicación) Bioprocessing
Esterilidad Estéril
Estilo de los tubos C-Flex
Descripción de la película Aegis 5-14
Detalle del puerto 3 ports
Cantidad 50 L