missing translation for 'onlineSavingsMsg'
Learn More

TIMP-2 Ab-5 Mouse Monoclonal Antibody, Epredia™

Código de producto. 12629916 Tienda Epredia Productos
Change view
Click to view available options
Cantidad:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugado:
Biotin
Unconjugated
Formulario:
Concentrated
Prediluted
Concentración:
1mg/mL
200μg/mL
Prediluted
Tamaño de la unidad:
1 ml
100 microlitros
200 microlitros
500 microlitros
7 ml
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad Conjugado Formulario Concentración unitSize
12629916 7 mL Unconjugated Prediluted Prediluted 7 ml
12629906 100 μL Biotin Concentrated 200μg/mL 100 microlitros
12639906 500 μL Biotin Concentrated 200μg/mL 500 microlitros
12649906 100 μL Unconjugated Concentrated 200μg/mL 100 microlitros
12679906 100 μL Unconjugated Concentrated 1mg/mL 100 microlitros
12699906 200 μL Unconjugated Concentrated 1mg/mL 200 microlitros
12669906 500 μL Unconjugated Concentrated 200μg/mL 500 microlitros
12659906 1 mL Unconjugated Concentrated 200μg/mL 1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 12629916 Proveedor Epredia N.º de proveedor MS1485R7

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Provide accurate, reproducible results with the Epredia™ TIMP-2 Ab-5, Mouse Monoclonal Antibody.

TIMP-1 and TIMP-2 are inhibitory enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP-2 shows the highest binding affinity to both the latent (pro) and active forms of 72kDa Type IV collagenase (also known as MMP-2 or gelatinase A). It also has affinity for the active form of 92kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMP-2 inhibits the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.

Host Species: Mouse

Clone: 3A4

Isotype: IgG2a/κ

Species Reactivity: Human, Mouse, Rat, Rabbit, Cow and Guinea Ppig. Others not known.

Epitope: N-terminal

Immunogen: A synthetic peptide from the N-terminal region of human TIMP-2 protein

Molecular Weight: 21kDa

Positive Control: Conditioned, serum-free medium from (TPA-treated) HFL-1 cells; Pancreatic or colon CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)
  • Western Blotting
  • Immunoprecipitation (Not suitable)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno TIMP-2 Ab-5
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Monoclonal
Clon 3A4
Concentración Prediluted
Conjugado Unconjugated
Especie del huésped Mouse
Inmunógeno A synthetic peptide from the N-terminal region of human TIMP-2 protein
Cantidad 7 mL
Estado normativo RUO
Disciplina de investigación Proteases And Inhibitors
Primario o secundario Primary
Especies diana Bovine, Guinea Pig, Human, Mouse, Rabbit, Rat
Formulario Prediluted
Isotype IgG2a κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.