Learn More
Thrombospondin (TSP) Ab-4 Mouse Monoclonal Antibody, Epredia™
Código de producto. p-4562863 Tienda Epredia Productos
Change view
Click to view available options
Cantidad:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugado:
Biotin
Unconjugated
Formulario:
Concentrated
Prediluted
Concentración:
1mg/mL
200μg/mL
Prediluted
Tamaño de la unidad:
1 ml
100 microlitros
200 microlitros
500 UL
500 microlitros
7 ml
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad Conjugado Formulario Concentración unitSize
12611397 100 μL Biotin Concentrated 200μg/mL 100 microlitros
12621397 500 μL Biotin Concentrated 200μg/mL 500 microlitros
12641397 100 μL Unconjugated Concentrated 200μg/mL 100 microlitros
12661397 100 μL Unconjugated Concentrated 1mg/mL 100 microlitros
12671397 200 μL Unconjugated Concentrated 1mg/mL 200 microlitros
12651397 500 μL Unconjugated Concentrated 200μg/mL 500 UL
12631397 1 mL Unconjugated Concentrated 200μg/mL 1 ml
12621407 7 mL Unconjugated Prediluted Prediluted 7 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Código de producto. 12611397 Proveedor Epredia N.º de proveedor MS421B0
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos
Provide accurate, reproducible results Epredia™ Thrombospondin (TSP) Ab-4, Mouse Monoclonal Antibody.
Thrombospondin is a protein from platelet a-granules. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.
Host Species: Mouse
Clone: A6.1
Isotype: IgG1
Species Reactivity: Human, Cow, Pig, Horse, Dog, Sheep, Mouse and Rat. Others not known.
Epitope: Collagen Type V-binding domain of TSP
Immunogen: Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets
Molecular Weight: ∽450kDa (nonreduced), 170 to 180kDa (reduced)
Positive Control: Tonsil or Placenta
Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix
Recommended for:
  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)
  • Electron Microscopy
  • Flow Cytometry
Note:
USA: RUO; Int'l: RUO
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Thrombospondin (TSP) Ab-4
Aplicaciones Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Clasificación Monoclonal
Clon A6.1
Concentración 200μg/mL
Conjugado Biotin
Especie del huésped Mouse
Inmunógeno Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets
Cantidad 100 μL
Estado normativo RUO
Disciplina de investigación Angiogenesis
Primario o secundario Primary
Especies diana Bovine, Canine, Equine, Human, Mouse, Pig, Rat, Sheep
Formulario Concentrated
Isotype IgG1