missing translation for 'onlineSavingsMsg'
Learn More

Thrombospondin (TSP) Ab-4 Mouse Monoclonal Antibody, Epredia™

Código de producto. 12661397 Tienda Epredia Productos
Change view
Click to view available options
Cantidad:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugado:
Biotin
Unconjugated
Formulario:
Concentrated
Prediluted
Concentración:
1mg/mL
200μg/mL
Prediluted
Tamaño de la unidad:
1 ml
100 microlitros
200 microlitros
500 UL
500 microlitros
7 ml
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad Conjugado Formulario Concentración unitSize
12661397 100 μL Unconjugated Concentrated 1mg/mL 100 microlitros
12611397 100 μL Biotin Concentrated 200μg/mL 100 microlitros
12621397 500 μL Biotin Concentrated 200μg/mL 500 microlitros
12641397 100 μL Unconjugated Concentrated 200μg/mL 100 microlitros
12671397 200 μL Unconjugated Concentrated 1mg/mL 200 microlitros
12651397 500 μL Unconjugated Concentrated 200μg/mL 500 UL
12631397 1 mL Unconjugated Concentrated 200μg/mL 1 ml
12621407 7 mL Unconjugated Prediluted Prediluted 7 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 12661397 Proveedor Epredia N.º de proveedor MS421P1ABX

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Provide accurate, reproducible results Epredia™ Thrombospondin (TSP) Ab-4, Mouse Monoclonal Antibody.

Thrombospondin is a protein from platelet a-granules. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.

Host Species: Mouse

Clone: A6.1

Isotype: IgG1

Species Reactivity: Human, Cow, Pig, Horse, Dog, Sheep, Mouse and Rat. Others not known.

Epitope: Collagen Type V-binding domain of TSP

Immunogen: Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets

Molecular Weight: ∽450kDa (nonreduced), 170 to 180kDa (reduced)

Positive Control: Tonsil or Placenta

Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)
  • Electron Microscopy
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Thrombospondin (TSP) Ab-4
Aplicaciones Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Clasificación Monoclonal
Clon A6.1
Concentración 1mg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Inmunógeno Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets
Cantidad 100 μL
Estado normativo RUO
Disciplina de investigación Angiogenesis
Primario o secundario Primary
Especies diana Bovine, Canine, Equine, Human, Mouse, Pig, Rat, Sheep
Formulario Concentrated
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.