missing translation for 'onlineSavingsMsg'
Learn More

Surfactant Protein B (Precursor) Ab-1 Mouse Monoclonal Antibody, Epredia™

Código de producto. 12622087 Tienda Epredia Productos
Change view
Click to view available options
Cantidad:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugado:
Biotin
Unconjugated
Formulario:
Concentrated
Prediluted
Concentración:
1mg/mL
200μg/mL
Prediluted
Tamaño de la unidad:
1 ml
100 microlitros
200 microlitros
500 UL
500 microlitros
7 ml
Código de producto. Cantidad Conjugado Formulario Concentración unitSize
12622087 200 μL Unconjugated Concentrated 1mg/mL 200 microlitros
12652077 100 μL Biotin Concentrated 200μg/mL 100 microlitros
12672077 500 μL Biotin Concentrated 200μg/mL 500 microlitros
12662077 1 mL Biotin Concentrated 200μg/mL 1 ml
12692077 100 μL Unconjugated Concentrated 200μg/mL 100 microlitros
12602087 100 μL Unconjugated Concentrated 1mg/mL 100 microlitros
12612087 500 μL Unconjugated Concentrated 200μg/mL 500 UL
12682077 1 mL Unconjugated Concentrated 200μg/mL 1 ml
12662087 7 mL Unconjugated Prediluted Prediluted 7 ml
9 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 12622087

Marca: Epredia MS704PABX

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Epredia™ Surfactant Protein B (Precursor) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Pulmonary surfactant is a complex mixture of phospholipids and proteins that is secreted from type II cells in alveoli and reduces the surface tension at the alveolar air-liquid interface, providing alveolar stability necessary for normal ventillation. Four distinct proteins isolated from pulmonary surfactant are termed surfactant proteins A, B, C, and D. SP-A (28-36kDa) and SP-D (43kDa) are collagenous carbohydrate-binding proteins, whereas SP-B (8-9kDa) and SP-C (4kDa) are non-collagenous hydrophobic proteins. SP-B is expressed in pulmonary adenocarcinomas with acinar, papillary, bronchioloalveolar, and solid growth patterns. Squamous cell and large cell carcinomas of the lung and nonpulmonary adenocarcinomas do not express SP-B. ProSP-B is glycosylated in the Golgi apparatus and undergoes carboxy- and amino-terminal proteolysis by a cathepsin D-like protease.

Host Species: Mouse

Clone: SPB01; same as 1-2-A5-9

Isotype: IgG2a/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant prosurfactant protein B

Molecular Weight: 42-46kDa

Positive Control: Lung adenocarcinoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Surfactant Protein B (Precursor) Ab-1
Aplicaciones Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon SPB01; same as 1-2-A5-9
Concentración 1mg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Inmunógeno Recombinant pro-surfactant protein B
Cantidad 200 μL
Estado normativo RUO
Disciplina de investigación Cancer and Tumor Biology
Primario o secundario Primary
Especies diana Human, Rat
Formulario Concentrated
Isotype IgG2a κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.