missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ SREBP-1 (Sterol Regulatory Element Binding Protein-1) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Código de producto. p-4560755 Tienda Epredia Productos
Change view
Click to view available options
Cantidad:
100 μL
200 μL
500 μL
1 mL
Concentración:
1mg/mL
200μg/mL
Tamaño de la unidad:
1 ml
100 microlitros
200 microlitros
500 UL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad Concentración unitSize
12689256 100 μL 200μg/mL 100 microlitros
12699256 500 μL 200μg/mL 500 UL
12669256 1 mL 200μg/mL 1 ml
12609266 100 μL 1mg/mL 100 microlitros
12619266 200 μL 1mg/mL 200 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 12689256 Proveedor Epredia™ N.º de proveedor MS1207P0

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Epredia™ SREBP-1 (Sterol Regulatory Element Binding Protein-1) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

SREBP-1 and -2 are transcription factors which participate in the control of cholesterol homeostasis. SREBPs proteins, which are attached to the endoplasmic reticulum and nuclear envelope, are proteolytically cleaved and thus activated in response to conditions of low cellular sterol. Upon activation of SREBP-1 or -2, an ∽480-500 amino acid, N-terminal cleavage fragment of these proteins enters the nucleus and activates transcription of enzymes and other proteins required for cholesterol synthesis. SCA (SREBP-cleavage activity) and caspase-3 proteases cleave SREBPs. SREBP proteins containing point mutations at caspase-3 cleavage sites (Asp460 in SREBP-1 and Asp468 in SREBP-2) do not become cleaved following induction of apoptosis, suggesting that SREBPs may play some role in apoptotic processes.

Host Species: Mouse

Clone: 2A4

Isotype: IgG1/κ

Species Reactivity: Human and Hamster. Others not known.

Epitope: aa 301-407

Immunogen: Recombinant protein encoding N-terminal amino acids 301-407 (the bHLH/leucine zipper domain) of human SREBP-1

Molecular Weight: 125kDa (precursor) and 60-70kDa (cleaved)

Positive Control: HeLa and IMR5 cells

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Not verified)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno SREBP-1 (Sterol Regulatory Element Binding Protein-1) Ab-1
Aplicaciones Immunoprecipitation, Western Blot
Clasificación Monoclonal
Clon 2A4
Concentración 200μg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Inmunógeno Recombinant protein encoding N-terminal amino acids 301-407 (the bHLH/leucine zipper domain) of human SREBP-1
Cantidad 100 μL
Estado normativo RUO
Disciplina de investigación Transcription Regulation
Primario o secundario Primary
Especies diana Hamster, Human
Formulario Concentrated
Isotype IgG2a κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.