Learn More
Epredia™ Lab Vision™ Placental Alkaline Phosphatase (PLAP) Rabbit Monoclonal Antibody,
Código de producto. p-4557286 Tienda Epredia Productos
Change view
Click to view available options
Cantidad:
100 μL
500 μL
1 mL
7 mL
12 mL
Tamaño de la unidad:
1 ml
100 microlitros
12 ml
500 microlitros
7 ml
12653737 100 μL 100 microlitros
12693737 500 μL 500 microlitros
12643737 1 mL 1 ml
12613737 7 mL 7 ml
12623737 12 mL 12 ml
5 options
Código de producto. 12653737
Marca: Epredia™ RM9115S0
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos
Placental Alk. Phosphatase (PLAP), Rabbit Monoclonal Ab is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal abs are more sensitive and specific than other available markers.
Reacts with a membrane-bound isoenzyme (Regan and Nagao type) of Placental Alkaline Phosphatase (PLAP) occurring in the placenta during the 3rd trimester of gestation. This antibody is highly specific to PLAP and shows no cross-reaction with other isoenzymes of alkaline phosphatases. It is useful in the identification of testicular germ cell tumors. Unlike germ cell tumors, PLAP-positive somatic cell tumors uniformly express epithelial membrane antigen (EMA).
Host Species: Rabbit
Clone: SP15
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: Recombinant protein encoding human Placental Alkaline Phosphatase (PLAP)
Molecular Weight: 60-70kDa
Positive Control: Placenta or seminoma
Cellular Localization: Membrane
Recommended for:
  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Placental Alk Phosphatase (PLAP)
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon SP15
Conjugado Unconjugated
Especie del huésped Rabbit
Inmunógeno Recombinant protein encoding human Placental Alkaline Phosphatase (PLAP)
Cantidad 100 μL
Estado normativo IVD
Disciplina de investigación Cancer and Tumor Biology
Primario o secundario Primary
Especies diana Human
Formulario Concentrated