missing translation for 'onlineSavingsMsg'
Learn More

Microphthalmia (Transcription Factor) (MiTF) Ab-2 Mouse Monoclonal Antibody, Epredia™

Código de producto. 12672307 Tienda Epredia Productos
Change view
Click to view available options
Cantidad:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugado:
Unconjugated
Formulario:
Concentrated
Prediluted
Concentración:
1mg/mL
200μg/mL
Tamaño de la unidad:
1 ml
100 microlitros
200 microlitros
500 microlitros
7 ml
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad Conjugado Formulario Concentración unitSize
12672307 100 μL Unconjugated Concentrated 1mg/mL 100 microlitros
12652307 100 μL Unconjugated Concentrated 200μg/mL 100 microlitros
12682307 200 μL Unconjugated Concentrated 1mg/mL 200 microlitros
12662307 500 μL Unconjugated Concentrated 200μg/mL 500 microlitros
12642307 1 mL Unconjugated Concentrated 200μg/mL 1 ml
12612317 7 mL Unconjugated Prediluted - 7 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 12672307 Proveedor Epredia N.º de proveedor MS772P1ABX

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Ensure accurate, reproducible results in immunohistochemistry experiments with the Epredia™ Microphthalmia (Transcription Factor) (MiTF) Ab-2, Mouse Monoclonal Antibody.

Mi is a basic helix-loop-helix-leucine zipper (b-HLH-ZIP) transcription factor implicated in pigmentation, mast cells and bone development. The mutation of Mi causes Waardenburg Syndrome type II in humans. In mice, a profound loss of pigmented cells in the skin eye and inner ear results, as well as osteopetrosis and defects in natural killer and mast cells. There are two known isoforms of Mi differing by 66 amino acids at the NH2 terminus. Shorter forms are expressed in melanocytes and run as two bands at 52kDa and 56kDa, while the longer Mi form runs as a cluster of bands at 60-70kDa in osteoclasts and in B16 melonoma cells (but not other melanoma cell lines), as well as mast cells and heart.

Host Species: Mouse

Clone: D5

Isotype: IgG1/κ

Species Reactivity: Human and Dog. Does not react with mouse and rat. Others not known.

Epitope: N-terminal

Immunogen: N-terminal fragment of human Mi protein

Molecular Weight: 52-56kDa doublet

Positive Control: 501 Mel cells, Melanoma

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Microphthalmia (Transcription Factor) (MiTF) Ab-2
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon D5
Concentración 1mg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Inmunógeno N-terminal fragment of human Mi protein
Cantidad 100 μL
Estado normativo RUO
Disciplina de investigación Transcription Regulation
Primario o secundario Primary
Especies diana Canine, Human
Formulario Concentrated
Isotype IgG2a κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.