Learn More
Microphthalmia (Transcription Factor) (MiTF) Ab-2 Mouse Monoclonal Antibody, Epredia™
Código de producto. 12662307 Tienda Epredia Productos
Change view
Click to view available options
Cantidad:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugado:
Unconjugated
Formulario:
Concentrated
Prediluted
Concentración:
1mg/mL
200μg/mL
Tamaño de la unidad:
1 ml
100 microlitros
200 microlitros
500 microlitros
7 ml
Código de producto. Cantidad Conjugado Formulario Concentración unitSize
12662307 500 μL Unconjugated Concentrated 200μg/mL 500 microlitros
12652307 100 μL Unconjugated Concentrated 200μg/mL 100 microlitros
12672307 100 μL Unconjugated Concentrated 1mg/mL 100 microlitros
12682307 200 μL Unconjugated Concentrated 1mg/mL 200 microlitros
12642307 1 mL Unconjugated Concentrated 200μg/mL 1 ml
12612317 7 mL Unconjugated Prediluted - 7 ml
6 options
Código de producto. 12662307
Marca: Epredia MS772P1
Ensure accurate, reproducible results in immunohistochemistry experiments with the Epredia™ Microphthalmia (Transcription Factor) (MiTF) Ab-2, Mouse Monoclonal Antibody.
Mi is a basic helix-loop-helix-leucine zipper (b-HLH-ZIP) transcription factor implicated in pigmentation, mast cells and bone development. The mutation of Mi causes Waardenburg Syndrome type II in humans. In mice, a profound loss of pigmented cells in the skin eye and inner ear results, as well as osteopetrosis and defects in natural killer and mast cells. There are two known isoforms of Mi differing by 66 amino acids at the NH2 terminus. Shorter forms are expressed in melanocytes and run as two bands at 52kDa and 56kDa, while the longer Mi form runs as a cluster of bands at 60-70kDa in osteoclasts and in B16 melonoma cells (but not other melanoma cell lines), as well as mast cells and heart.
Host Species: Mouse
Clone: D5
Isotype: IgG1/κ
Species Reactivity: Human and Dog. Does not react with mouse and rat. Others not known.
Epitope: N-terminal
Immunogen: N-terminal fragment of human Mi protein
Molecular Weight: 52-56kDa doublet
Positive Control: 501 Mel cells, Melanoma
Cellular Localization: Nuclear
Recommended for:
  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Gel Supershift
Note:
USA: RUO; Int'l: RUO
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Microphthalmia (Transcription Factor) (MiTF) Ab-2
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon D5
Concentración 200μg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Inmunógeno N-terminal fragment of human Mi protein
Cantidad 500 μL
Estado normativo RUO
Disciplina de investigación Transcription Regulation
Primario o secundario Primary
Especies diana Canine, Human
Formulario Concentrated
Isotype IgG2a κ