Learn More
Interferon-α (II)/IFN-α (II) Mouse Monoclonal Antibody, Epredia™
Epredia™ Interferon-α (II)/IFN-α (II), Mouse Monoclonal Antibody provides accurate, reproducible results.
Marca: Epredia MS-574-P1ABX
Descripción
The IFN-α family consists of 24 or more genes or pseudo-genes. IFN-αII) is one of the two distinct families (I and II) of human IFN-α. The α interferons are mainly produced by lymphocytes, monocytes, macrophages, and cell lines such as Namalwa and KG1 following induction by viruses, nucleic acids, and glucocorticoid hormones. They are involved in virus resistance on target cells, inhibition of cell proliferation, induction of cytokines and regulation of expression of MHC class I antigens.Host Species: Mouse
Clone: N27
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: aa 43-53
Immunogen: Purified recombinant IFN-α 2c
Molecular Weight: 16-27kDa
Positive Control: Stimulated human peripheral blood mononuclear cells
Recommended for:
- Blocking study
Note:
USA: RUO; Int'l: RUO
Especificaciones
| Interferon-alpha (II)/IFN-alpha (II) | |
| N27 | |
| Unconjugated | |
| Purified recombinant IFN-alpha 2c | |
| RUO | |
| Primary | |
| Concentrated |
| Monoclonal | |
| 1mg/mL | |
| Mouse | |
| 100 μL | |
| Cytokines | |
| Human | |
| IgG2a κ |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.