missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
The type I keratin 17 (K17) shows a peculiar localization in human epithelial appendages including hair follicles, which undergo a growth cycle throughout adult life. Additionally K17 is induced, along with K6 and K16, early after acute injury to human skin. Predominant expression of K17 and the frequent expression of K8 and K19, with little K6/K16 and K1/K10 expression are the characteristic features of basal cell carcinomas (BCC), suggesting that BCC is differentiated towards undifferentiated follicular epithelia, most probably hair bulge cells.
Host Species: Mouse
Clone: E3
Isotype: IgG2b/κ
Species Reactivity: Human and Rat. Others not known.
Epitope: Not determined
Immunogen: Cytokeratin preparation from rat enterocytes
Molecular Weight: 46 kDa
Positive Control: Squamous cell CA, Skin
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Especificaciones
Especificaciones
| Antígeno | Cytokeratin 17 Ab-1 |
| Aplicaciones | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | E3 |
| Conjugado | Unconjugated |
| Especie del huésped | Mouse |
| Inmunógeno | Keratin preparation from rat enterocytes |
| Método de purificación | Squamous cell CA, skin |
| Cantidad | 1 mL |
| Estado normativo | IVD |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?