missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Chorionic Gonadotropin Human/hCG Ab-5 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Tienda Epredia ProductosDescripción
Human Chorionic Gonadotropin (HCG) is a glycoprotein that is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the α- and Β-subunits. The α-subunit of HCG is nearly identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH). A germ cell tumor which is positive for cytokeratin, placental alkaline phosphatase (PLAP), and HCG but negative for EMA and AFP is probably a choriocarcinoma.
Host Species: Mouse
Clone: CG04 + CG05
Isotype: IgG2a/κ + IgG1/κ
Species Reactivity: Human. Others not known
Immunogen: Purified β-subunit of hCG
Molecular Weight: 22 kDa (β-hCG)
Positive Control: Placenta or chorioca
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting (Not optimum)
Note:
USA: RUO; Int'l: RUO
Especificaciones
Especificaciones
| Antígeno | Chorionic Gonadotropin, Human/hCG Ab-5 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | CG04 + CG05 |
| Concentración | 1mg/mL |
| Conjugado | Unconjugated |
| Especie del huésped | Mouse |
| Inmunógeno | Purified Beta-subunit of HCG |
| Cantidad | 100 μL |
| Estado normativo | RUO |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?