missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ CDC25A Ab-1 Mouse Monoclonal Antibody, 200µg/mL, BSA and azide
Tienda Epredia ProductosDescripción
Cyclin-dependent kinases are positively regulated by association with cyclins and negatively regulated by binding to inhibitory subunits. The activity of cyclin-dependent kinases is also regulated by the phosphorylation status, which is controlled by the antagonistic action of wee1 kinase and CDC25 phosphatases. Three CDC25 genes are present in human cells: CDC25A, CDC25B, and CDC25C. These three genes function at different phases of the cell cycle. Whereas CDC25A and CDC25B are expressed throughout the cell cycle, with peak expression in G1 for CDC25A and in both G1-S-phase and G2 for CDC25B, CDC25C is predominantly expressed in G2.
Host Species: Mouse
Clone: DCS-120
Isotype: IgG2a
Species Reactivity: Human and Rat. Others not known.
Immunogen: Purified recombinant CDC25A
Molecular Weight: 65 kDa
Positive Control: LS174T cells, Tonsil
Cellular Localization: Nuclear membrane
Recommended for:
- Western Blotting
- Immunoprecipitation (Native verified)
Note:
USA: RUO; Int'l: RUO
Especificaciones
Especificaciones
| Antígeno | CDC25A Ab-1 |
| Aplicaciones | Immunoprecipitation |
| Clasificación | Monoclonal |
| Clon | DCS-120 |
| Concentración | 200μg/mL |
| Conjugado | Unconjugated |
| Especie del huésped | Mouse |
| Inmunógeno | Purified recombinant CDC25A |
| Cantidad | 500 μL |
| Estado normativo | RUO |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?