Learn More
Amyloid Precursor Protein (APP) Rabbit Polyclonal Antibody, Epredia™
Código de producto. 12663337 Tienda Epredia Productos
Change view
Click to view available options
Cantidad:
100 μL
500 μL
1 mL
7 mL
Formulario:
Concentrated
Prediluted
Tamaño de la unidad:
1 ml
100 microlitros
500 UL
7 ml
Código de producto. Cantidad Formulario unitSize
12663337 100 μL Concentrated 100 microlitros
12633337 500 μL Concentrated 500 UL
12623337 1 mL Concentrated 1 ml
12673337 7 mL Prediluted 7 ml
4 options
Código de producto. 12663337
Marca: Epredia RB9023P0
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos
Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments.
Recognizes APP (b-APP770), MW 110kDa. Predicted to recognize Β-APP695 and Β-APP751. Amyloid precursor protein (APP) and APP-like proteins are transmembrane glycoproteins with a similar modular domain structure.
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse and Rat. Others not tested.
Epitope: N-terminal
Immunogen: A synthetic peptide corresponding to N-terminal of human APP
Molecular Weight: 110kDa
Positive Control: Brain
Cellular Localization: Cytoplasmic and extracellular
This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues
Recommended for:
  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Amyloid Precursor Protein (APP)
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 200µg/mL
Conjugado Unconjugated
Especie del huésped Rabbit
Inmunógeno A synthetic peptide corresponding to N-terminal of human APP
Cantidad 100 μL
Estado normativo IVD
Disciplina de investigación Neuroscience
Primario o secundario Primary
Especies diana Human, Mouse
Formulario Concentrated
Isotype IgG