Learn More
Amyloid Precursor Protein (APP) Rabbit Polyclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments.
Marca: Epredia RB-9023-P
Descripción
Recognizes APP (b-APP770), MW 110kDa. Predicted to recognize Β-APP695 and Β-APP751. Amyloid precursor protein (APP) and APP-like proteins are transmembrane glycoproteins with a similar modular domain structure.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse and Rat. Others not tested.
Epitope: N-terminal
Immunogen: A synthetic peptide corresponding to N-terminal of human APP
Molecular Weight: 110kDa
Positive Control: Brain
Cellular Localization: Cytoplasmic and extracellular
This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Especificaciones
| Amyloid Precursor Protein (APP) | |
| Polyclonal | |
| Rabbit | |
| 1 mL | |
| Neuroscience | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| Unconjugated | |
| A synthetic peptide corresponding to N-terminal of human APP | |
| IVD | |
| Primary | |
| Concentrated |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.