missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Theanine, MedChemExpress
Description
L-Theanine (L-Glutamic Acid γ-ethyl amide)is a non-protein amino acid contained in green tea leaves, which blocks the binding of L-glutamic acid to glutamate receptors in the brain, and with neuroprotective and anti-oxidative activities. L-Theanine causes anti-stress effects via the inhibition of cortical neuron excitation by oral intake.
Specifications
Specifications
| Nombre del producto químico o material | L-Theanine |
| Forma física | Solid |
| CAS | 3081-61-6 |
| Color | Blanco |
| Cantidad | 200 mg |
| Notas de grado de pureza | Research |
| Fórmula molecular | C7H14N2O3 |
| Para utilizar con (aplicación) | Neuroscience-Neurodegeneration |
| Sinónimo | L-Glutamic Acid γ-ethyl amide Nγ-Ethyl-L-glutamine |
| Información de solubilidad | H2O : 150 mg/mL (861.08 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?