Learn More
Tocris Bioscience™ KYL
Descripción
KYL is an EphA4 receptor tyrosine kinase inhibitor (Kd = 0.8 μM); inhibits EphA4-EphrinA5 interactions (IC50 = 6.34 μM). Prevents AβO induced synaptic damage, dendritic spine loss and prevents the blocking of LTP in hippocampal CA3-CA1 transmissions. Exhibits a long half life in cell culture media (8 and 12 hours in PC3 and C2C12 media respectively). Neuroprotective.
Especificaciones
Especificaciones
| Descripción | KYL |
| Peso molecular | 1465.75 |
| Tipo de producto | KYL |
| Cantidad | 1 mg |
| Secuencia | Lys-Tyr-Leu-Pro-Tyr-Trp-Pro-Val-Leu-Ser-Ser-Leu |
Para uso exclusivo en investigación