missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KVbeta2 (KCNAB2) Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15887103
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
15887103 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15887103

Marca: Invitrogen™ PA568695

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

Kcnab2 is a voltage-gated potassium channels (KV) belong to the 6-TM family of potassium channel that also comprises the Ca2+-activated Slo (actually 7-TM) and the Ca2+-activated SK subfamilies. The alpha-subunits contain a single pore-forming region and combine to form tetramers. Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. The diverse functions of potassium channels include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog(s). Kcnab2 is one of the beta subunits, which are auxiliary proteins associating with functional Kv-alpha subunits. Alternative splicing of the Kcnab2 gene results in multiple transcript variants encoding distinct isoforms. Diseases associated with KCNAB2 include Chromosome 1P36 Deletion Syndrome.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno KVbeta2 (KCNAB2)
Aplicaciones Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 2% sucrose and 0.09% sodium azide
génica KCNAB2
N.º de referencia del gen Q13303
Alias de gen AKR6A5; Ckbeta2; F5; HKvbeta2; HKvbeta2.1; HKvbeta2.2; I2rf5; K(+) channel subunit beta-2; KCNA2B; KCNAB2; KCNAB2.1; KCNAB2.2; Kcnb3; KCNK2; KVB2; Kvbeta2; Kv-beta-2; Kvbeta2.1; Kvbeta2.2; neuroimmune protein F5; potassium channel; potassium channel Kv beta 2.1 subunit; potassium channel Kv beta 2.2 subunit; potassium channel, voltage gated shaker-related subfamily A regulatory beta subunit 2; potassium channel, voltage gated subfamily A regulatory beta subunit 2; potassium channel, voltage-gated shaker-related subfamily A regulatory beta subunit 2; potassium voltage gated channel, shaker related subfamily, beta member 2; potassium voltage-gated channel subfamily A regulatory beta subunit 2; potassium voltage-gated channel, shaker-related subfamily, beta member 2; potassium voltage-gated channel, shaker-related subfamily, beta member 2, transcript variant 1; potassium voltage-gated channel, shaker-related subfamily, beta member 2, transcript variant 2; voltage-gated potassium channel subunit beta-2
Símbolos de los genes KCNAB2
Especie del huésped Rabbit
Inmunógeno synthetic peptide directed towards the middle region of human KCNAB2.
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 8514
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.