missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: GSQISEENTK LRRQSGFSVA GKDKSPKKAS ENAKDSSLSP SGESQLRARQ LALLREVEMN WYLKLCDLSS EHTTVCTTGM Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 81%.
Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. The KCNAB1 gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily.
Especificaciones
Especificaciones
| Antígeno | KVbeta1 (KCNAB1) |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | KCNAB1 |
| N.º de referencia del gen | Q14722 |
| Alias de gen | AKR6A3; Akr8a8; hKvb3; hKvBeta3; K(+) channel subunit beta-1; K+ channel Beta1a chain; KCNA1B; KCNAB1; Kvb1; Kvb1.3; KVBETA1; Kv-beta-1; Kvbeta1.1; KvBeta3; mKv(beta)1; potassium channel beta 3 chain; potassium channel beta3 subunit; potassium channel shaker chain beta 1a; potassium channel subunit Kv beta 1; potassium channel subunit Kv beta 1.1; potassium channel subunit Kv beta 1.2; potassium channel subunit Kv beta 1.3; potassium channel, voltage gated subfamily A regulatory beta subunit 1; potassium channel, voltage-gated shaker related subfamily A regulatory beta subunit 1; potassium voltage gated channel shaker related subfamily beta member 1; potassium voltage gated channel, shaker related subfamily, beta member 1; potassium voltage-gated channel shaker-related subfamily beta member 1; potassium voltage-gated channel subfamily A member regulatory beta subunit 1; potassium voltage-gated channel, shaker-related subfamily, beta member 1; voltage-gated potassium channel beta subunit; voltage-gated potassium channel subunit beta-1 |
| Símbolos de los genes | KCNAB1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
