missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kv4.3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-87707
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Kv4.3 Polyclonal specifically detects Kv4.3 in Human, Rat samples. It is validated for Western Blot.
Especificaciones
| Kv4.3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3752 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| KCND3L, KCND3S, KSHIVB, Kv4.3, MGC142035, MGC142037, potassium ionic channel Kv4.3, potassium voltage-gated channel subfamily D member 3, potassium voltage-gated channel, Shal-related subfamily, member 3, sha1-related potassium channel Kv4.3, voltage-gated K+ channel, voltage-gated potassium channel Kv4.3, Voltage-gated potassium channel subunit Kv4.3 | |
| The immunogen is a synthetic peptide directed towards the middle region of human Kv4.3 | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido