missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KV1.8 (KCNA10) Polyclonal Antibody

Código de producto. 15564572
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15564572 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15564572 Proveedor Invitrogen™ N.º de proveedor PA541031

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Peptide sequence: DVCGWKEMEV ALVNFDNSDE IQEEPGYATD FDSTSPKGRP GGSSFSNGKI Sequence homology: Cow: 92%; Dog: 79%; Guinea Pig: 93%; Horse: 85%; Human: 100%; Mouse: 86%; Rat: 93%.

KCNA10 is a member of the potassium channel, voltage-gated, shaker-related subfamily. This member contains six membrane-spanning domains with a shaker-type repeat in the fourth segment. It is specifically regulated by cGMP and postulated to mediate the effects of substances that increase intracellular cGMP. This gene is intronless, and the gene is clustered with genes KCNA2 and KCNA3 on chromosome 1.Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog(s). This gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily. This member contains six membrane-spanning domains with a shaker-type repeat in the fourth segment. It is specifically regulated by cGMP and postulated to mediate the effects of substances that increase intracellular cGMP. This gene is intronless, and the gene is clustered with genes KCNA2 and KCNA3 on chromosome 1.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno KV1.8 (KCNA10)
Aplicaciones Immunohistochemistry, Western Blot
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 2% sucrose and 0.09% sodium azide
génica KCNA10
N.º de referencia del gen Q16322
Alias de gen cyclic GMP gated potassium channel; Gm1962; Kcn1; KCNA10; Kcna8; Kv1.8; potassium channel, voltage gated shaker related subfamily A, member 10; potassium voltage gated channel, shaker related subfamily, member 8; potassium voltage-gated channel subfamily A member 10; potassium voltage-gated channel, shaker-related subfamily, member 10; potassium voltage-gated channel, shaker-related subfamily, member 8; voltage-gated potassium channel subunit Kv1.8
Símbolos de los genes KCNA10
Especie del huésped Rabbit
Inmunógeno Synthetic peptide directed towards the N-terminal of human KCNA10 (aa 2-51).
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3744
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.