missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KV1.6 (KCNA6) Polyclonal Antibody
GREENER_CHOICE

Código de producto. 17298263 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17298263 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17298263 Proveedor Invitrogen™ N.º de proveedor PA5115487

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Kv1.6.

Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog. This gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily. This member contains six membrane-spanning domains with a shaker-type repeat in the fourth segment. It belongs to the delayed rectifier class. The coding region of this gene is intronless, and the gene is clustered with genes KCNA1 and KCNA5 on chromosome 12.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno KV1.6 (KCNA6)
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide
génica KCNA6
N.º de referencia del gen P17658, P17659, Q61923
Alias de gen FLJ25134; HBK2; human brain potassium channel-2; KCNA6; KV1.6; kv2; MK1.6; potassium channel, voltage gated shaker related subfamily A, member 6; potassium channel-Kv2; potassium voltage gated channel, shaker related subfamily, member 6; potassium voltage gated channel, shaker related, subfamily, member 6; potassium voltage-gated channel subfamily A member 6; potassium voltage-gated channel, shaker-related subfamily, member 6; potassium voltage-gated channel, shaker-related, subfamily, member 6; PPP1R96; protein phosphatase 1, regulatory subunit 96; RCK2; voltage-gated potassium channel HBK2; voltage-gated potassium channel protein Kv1.6; voltage-gated potassium channel subunit Kv1.6; voltage-gated potassium channel subunit Kv2
Símbolos de los genes KCNA6
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from human KCNA6(Accession P17658), corresponding to amino acid residues R490-V529.
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 16494, 3742, 64358
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.