Learn More
Invitrogen™ KV1.2 (KCNA2) Polyclonal Antibody

Descripción
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Immunogen sequence is identical to the related mouse and rat sequences. Positive Control - WB: Rat Kidney Tissue, Rat Brain Tissue, Mouse Brain Tissue, Mouse Kidney Tissue. ICC/IF: U20S cell. Flow: A431 cell, C6 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Especificaciones
Especificaciones
| Antígeno | KV1.2 (KCNA2) |
| Aplicaciones | Flow Cytometry, Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 500 μg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 5mg BSA and 0.05mg sodium azide |
| génica | KCNA2 |
| N.º de referencia del gen | P16389, P63141, P63142 |
| Alias de gen | Akr6a4; BK2; CSMK1; delayed rectifier K+ channel; EIEE32; ENSMUSG00000074335; Gm10672; HBK5; HK4; HUKIV; k(v)1.2; KC22; Kca1-2; kcna2; kcna2.S; kcna2-a; KV1.2; Kv1.2' potassium channel; LOC100537815; MK2; Mk-2; NGK1; Potassium (K+) channel protein alpha 2, voltage dependent; potassium channel (BGK5); potassium channel subunit Kv 1.2; potassium channel, voltage gated shaker related subfamily A, member 2; potassium channel, voltage gated shaker related subfamily A, member 2 S homeolog; potassium voltage gated channel shaker related subfamily member 2; potassium voltage-gated channel subfamily A member 2; potassium voltage-gated channel, shaker-related subfamily, member 2; potassium voltage-gated channel, shaker-related subfamily, member 2 a; RAK; RBK2; RCK5; RP11-284N8.1; voltage-dependent K channel; voltage-gated channel, shaker-related subfamily, member 2; voltage-gated K(+) channel HuKIV; Voltage-gated potassium channel HBK5; voltage-gated potassium channel isoform 2; voltage-gated potassium channel isoform 3; voltage-gated potassium channel protein Kv1.2; Voltage-gated potassium channel subunit Kv1.2; XELAEV_18011981mg; XSha2 |
| Símbolos de los genes | KCNA2 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.