Learn More
Invitrogen™ KV1.1 (KCNA1) Polyclonal Antibody
GREENER_CHOICE
Código de producto. 11536042
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
11536042 100 μg 100 microgramos
1 options
Código de producto. 11536042
Marca: Invitrogen™ PA112652
Rabbit Polyclonal Antibody
A suggested positive control for this product is rat brain lysate.
This gene encodes a voltage-gated delayed potassium channel that is phylogenetically related to the Drosophila Shaker channel. The encoded protein has six putative transmembrane segments, and the loop between S5 and S6 forms the pore and contains the conserved selectivity filter motif. The functional channel is a homotetramer. The N-terminus of the channel is associated with beta subunits that can modify the inactivation properties of the channel as well as affect expression levels. The C-terminus of the channel is complexed to a PDZ domain protein that is responsible for channel targeting. Mutations in this gene have been associated with myokymia with periodic ataxia.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno KV1.1 (KCNA1)
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.08% sodium azide
génica Kcna1
N.º de referencia del gen P10499, P16388, Q09470
Alias de gen AEMK; AI840627; brain potassium channel protein-1; EA1; HBK1; HUK1; IA; Kca1-1; Kcna; Kcna1; Kcpvd; Kv1.1; MBK1; mceph; megencephaly; MGC126782; MGC138385; MK1; Mk-1; MKI; potassium (K+) channel protein voltage dependent; potassium channel, voltage gated shaker related subfamily A, member 1; potassium voltage gated channel shaker related subfamily member 1; potassium voltage gated channel, shaker related subfamily, member 1; potassium voltage-gated channel subfamily A member 1; potassium voltage-gated channel, shaker-related subfamily, member 1; potassium voltage-gated channel, shaker-related subfamily, member 1 (episodic ataxia with myokymia); RBK1; RBKI; RCK1; Shak; Voltage-gated K(+) channel HuKI; voltage-gated potassium channel HBK1; Voltage-gated potassium channel subunit Kv1.1
Símbolos de los genes Kcna1
Especie del huésped Rabbit
Inmunógeno Synthetic peptide, conjugated to KLH, corresponding to the rat Kv1.1 potassium channel.
Método de purificación Ammonium sulfate precipitation
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 16485, 24520, 3736
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG