missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KTELC1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24409-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
KTELC1 also termed as CLP46 is a glucosyltransferase, which implicates in Protein modification and glycosylation. KTELC1 encodes a 392 amino acids protein and its molecular weight is 46 kDa, but is exists glycosylation modification. KTELC1 is localized in endoplasmic reticulum lumen and abundantly expressed in liver, brain, heart, skeletal muscle, spleen, kidney, placenta, lung and peripheral blood leukocyte. KTELC1 may regulation in cell proliferation and Notch signal pathway.Especificaciones
| KTELC1 | |
| Polyclonal | |
| Unconjugated | |
| POGLUT1 | |
| C3orf9, CAP10 like 46 kDa protein, CLP46, hCLP46, KDELCL1, KTELC1, MDS010, MDSRP, UNQ490/PRO1006 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56983 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8NBL1 | |
| POGLUT1 | |
| KTELC1 Fusion Protein Ag19942 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido