missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT35 Monoclonal Antibody (OTI1A11), TrueMAB™, OriGene
Descripción
KRT35 Monoclonal antibody specifically detects KRT35 in Human samples. It is validated for Western Blot
Especificaciones
Especificaciones
| Antígeno | KRT35 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI1A11 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | KIF6 |
| N.º de referencia del gen | Q6ZMV9 |
| Alias de gen | AI662400; Ha5; Ha-5; hair keratin; hair keratin acidic 5; hair keratin, type I Ha5; hard keratin, type I, 5; HHA5; HHa5 hair keratin type I intermediate filament; HKA5; K35; Ka30; keratin 35; keratin 35, type I; keratin complex 1, acidic, gene 2; keratin complex 1, acidic, gene 24; keratin complex 1, gene 24; keratin, hair, acidic, 5; keratin, type I cuticular Ha5; Keratin-35; KRT hHa5; Krt1-2; Krt1-24; KRT35; KRTHA5; type I hair keratin KA30; Unknown (protein for MGC:139866); wool (hair) keratin intermediate filament protein |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?