missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT19 Mouse anti-Human, Clone: UMAB186, lyophilized, UltraMAB™
Descripción
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Cytokeratin 19 is part of a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight from 40-68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial maligcies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. For example, cytokeratin 19 is not expressed in hepatocytes, therefore, it is useful in the identification of liver metastasis.
Especificaciones
Especificaciones
| Antígeno | KRT19 |
| Aplicaciones | Immunohistochemistry (Paraffin), Peptide Array |
| Clasificación | Monoclonal |
| Clon | UMAB186 |
| Conjugado | Unconjugated |
| Formulación | PBS with 8% trehalose and no preservative; pH 7.3 |
| génica | KRT19 |
| N.º de referencia del gen | P08727 |
| Alias de gen | CK19, K19, K1CS |
| Símbolos de los genes | KRT19 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?