missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KNK437, MedChemExpress
Description
KNK437 is a HSP inhibitor, and inhibits the induction of HSP105, HSP70, and HSP40.
Specifications
Specifications
| Nombre del producto químico o material | KNK437 |
| CAS | 218924-25-5 |
| Color | Light Yellow |
| Notas de grado de pureza | Research |
| Fórmula molecular | C13H11NO4 |
| Cantidad | 200 mg |
| Sinónimo | Heat Shock Protein Inhibitor I |
| Información de solubilidad | DMSO : 31.25 mg/mL (127.43 mM; Need ultrasonic) |
| SMILES | O=CN1C(/C(CC1)=C/C2=CC=C(OCO3)C3=C2)=O |
| Peso molecular (g/mol) | 245.23 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?