missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLRG1 Antibody (2F1), DyLight 550, Novus Biologicals™
Hamster Monoclonal Antibody
Marca: Novus Biologicals FAB69441L
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
KLRG1 Monoclonal antibody specifically detects KLRG1 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Especificaciones
| KLRG1 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Hamster | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 2F1 | |
| Flow Cytometry, CyTOF-ready | |
| 2F1, CLEC15AMAFA-LIKE, C-type lectin domain family 15 member A, C-type lectin domain family 15, member A, ITIM-containing receptor MAFA-L, killer cell lectin-like receptor subfamily G member 1, killer cell lectin-like receptor subfamily G, member 1, MAFAL, MAFA-L, MAFA-like receptor, MAFAMAFA-2F1, Mast cell function-associated antigen, mast cell function-associated antigen (ITIM-containing), MGC13600 | |
| IL-2 activated NK cells from C57/BL6 mice, Accession # O88713 | |
| 0.1 mL | |
| Immunology, Innate Immunity, Signal Transduction | |
| 10219 | |
| Store at 4°C in the dark. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido