missing translation for 'onlineSavingsMsg'
Learn More

KLK8 Mouse anti-Human, Clone: OTI6D5, lyophilized, TrueMAB™

Código de producto. 17645161
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
17645161 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17645161

Marca: Origene Technologies CF506122

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo requiere una actualización de precios antes de ser solicitado.

Solicite el precio de este artículo aquí.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in tandem in a gene cluster on chromosome 19. The encoded protein may be involved in proteolytic cascade in the skin and may serve as a biomarker for ovarian cancer. Alternate splicing of this gene results in multiple transcript variants encoding different isoforms.

Specifications

Antígeno KLK8
Aplicaciones Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Clasificación Monoclonal
Clon OTI6D5
Conjugado Unconjugated
Formulación PBS with 8% trehalose and no preservative; pH 7.3
génica KLK8
N.º de referencia del gen O60259
Alias de gen HNP, NP, NRPN, PRSS19, TADG14
Símbolos de los genes KLK8
Especie del huésped Mouse
Inmunógeno Full length human recombit protein of human KLK8 produced in HEK293T cell.
Método de purificación Affinity Chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 11202
Especies diana Human
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.