missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: MNQVHVGGSL AVLGGKLYVS GGYDNTFELS DVVEAYDPET RAWSVVGRLP EPTFWHGSVS IFRQFMPQTF SGGRGFELD Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
Substrate-specific adapter of a BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex required for efficient chromosome alignment and cytokinesis. The BCR(KLHL21) E3 ubiquitin ligase complex regulates localization of the chromosomal passenger complex (CPC) from chromosomes to the spindle midzone in anaphase and mediates the ubiquitination of AURKB. Ubiquitination of AURKB by BCR(KLHL21) E3 ubiquitin ligase complex may not lead to its degradation by the proteasome.
Specifications
Specifications
| Antígeno | KLHL21 |
| Aplicaciones | Immunohistochemistry |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | KLHL21 |
| N.º de referencia del gen | Q9UJP4 |
| Alias de gen | KIAA0469; MGC99635; KLHL21 |
| Símbolos de los genes | KLHL21 |
| Especie del huésped | Rabbit |
| Show More |
Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?