Descripción
The KLF2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KLF2. This antibody recognizes human, mouse antigen. The KLF2 antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
Especificaciones
| Antígeno | KLF2 |
| Aplicaciones | Western Blot, ELISA |
| Concentración | 1000 μg/mL |
| Símbolos de los genes | KLF2, Krueppel like factor 2, Kruppel like factor 2 (lung), LKLF, Lung krueppel like factor |
| Especie del huésped | Rabbit |
| Cantidad | 150 μL |
| Primario o secundario | Primary |
| ID de gen (Entrez) | 10365 |
| Especies diana | Human, Mouse |
| Contenido y almacenamiento | Store at -20°C. Stable for one year after shipment. |
| Mostrar más |