missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Kir2.1 (KCNJ2) Polyclonal Antibody

Descripción
Antibody detects endogenous levels of total KCNJ2.
The KIR2 subunit family includes 2. 1, 2. 2, 2. 3 and 2. 4. Unlike G protein-coupled KIR3 subunits, KIR2. 1 requires both phosphorylation by PKA and ATP hydrolysis for functional activity. KIR2. 1 is expressed in the superior and inferior collicula and the pontine region of the brain, where it moderates synaptic transmission, like many other potassium channels. In the placenta, KIR2. 1 is expressed throughout gestation in cytotrophoblast cells. In the kidney, KIR2. 1 colocalizes with KIR5. 1 in the proximal tubule. KIR2. 1, 2. 2 and 2. 3 associate with the membrane-associated guanylate kinase synapseassociated protein 97 in the cerebellum and heart. Phosphorylation of KIR2. 2 by protein kinase A inhibits the associates with SAP97.
Especificaciones
Especificaciones
| Antígeno | Kir2.1 (KCNJ2) |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | Kcnj2 |
| N.º de referencia del gen | P63252 |
| Alias de gen | ATFB9; cardiac inward rectifier KIR2.1; cardiac inward rectifier potassium channel; EGK_08925; HHBIRK1; HHIRK 1; HHIRK1; HIRK 1; hIRK1; Inward rectifier K; inward rectifier K(+) channel Kir2.1; inward rectifier K+ channel KIR2.1; inward rectifier potassium channel; inward rectifier potassium channel 2; inwardly rectifying potassium channel Kir2.1; IRK 1; IRK1; IRK-1; IRK2; Kcnf1; KCNJ2; KIR2.1; LQT 7; LQT7; Potassium channel, inwardly rectifying subfamily J member 2; potassium channel, inwardly rectifying subfamily J, member 2; potassium inwardly rectifying channel subfamily J member 2; potassium inwardly-rectifying channel, subfamily J, member 2; potassium voltage-gated channel subfamily J member 2; RBL-IRK1; SQT 3; SQT3 |
| Símbolos de los genes | Kcnj2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?