Learn More
KIF5C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25897-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Kinesins are a superfamily of microtubule-associated motor proteins involved in a variety of cellular processes including membranous organelle transport and cell division. Kinesin has been found in a variety of organisms and cell types and is subject to spatial and temporal regulation. These proteins have a modular structure including a conserved motor domain of approximately 350 amino acids, which is responsible for microtubule binding and ATP hydrolysis. In addition to the motor domain, subfamily members share common domain organization, exhibit sequence similarity, motility properties, and cellular functions outside of the motor domai. There are currently three known Kinesin 5 family members denoted as A, B, and C. Kinesin 5A and kinesin 5C appear to be exclusively neuronal, whereas kinesin 5B appears to be ubiquitous in its expression.Especificaciones
| KIF5C | |
| Polyclonal | |
| Unconjugated | |
| KIF5C | |
| KIAA0531, KIF5C, kinesin family member 5C, Kinesin heavy chain isoform 5C, KINN, NKHC, NKHC 2, NKHC2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16574, 311024, 3800 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.79 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O60282, P28738, P56536 | |
| Kif5c | |
| KIF5C Fusion Protein Ag23163 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |