missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KIF2A Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Marca:  Invitrogen™ PA595751

Código de producto. 16568591

  • 530.00€ / 100 microlitros

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

Immunogen sequence: TSLNEDNESV TVEWIENGDT KGKEIDLESI FSLNPDLVPD EEIEPSPETP PPPASSAKVN KIVKNRRTVA SIKNDPPSRD NRVVGSARAR PSQFPEQSSS AQQNGSVSDI SPVQAAKKEF GPPSRRKSNC VKEVEKLQEK REKRRLQQQE LREKRAQDVD ATNPNYEIMC MIRDFRGSLD YRPLTTADPI D; Positive Samples: Rat brain; Cellular Location: Cytoplasm, centrosome, cytoskeleton, microtubule organizing center, spindle, spindle pole.

KIF2A, a gene encoding the kinesin family member 2A protein, plays a pivotal role in neuronal development and function. This gene is part of the kinesin-13 subfamily, characterized by its unique ability to depolymerize microtubules, which are essential components of the cytoskeleton responsible for maintaining cell shape, enabling motility, and facilitating intracellular transport. KIF2A is particularly significant in neural settings, where it regulates microtubule dynamics crucial for neuronal migration and dendritic development. Dysfunctional KIF2A, due to mutations, has been linked to early-onset neurodegenerative diseases such as progressive microcephaly and mental disabilities due to impaired microtubule regulation, reflecting its foundational role in correct brain development and architecture. Additionally, KIF2A is involved in spindle assembly and chromosome segregation during cell division, further cementing its importance in both neurodevelopment and general cellular functions.
TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

KIF2A
Polyclonal
Unconjugated
Kif2a
C530030B14Rik; CDCBM3; HK2; hypothetical protein; KIF2; kif2a; kif2-a; kif2a {ECO:0000250; kif2a.L; kinesin family member 2A; kinesin heavy chain family, member 2; kinesin heavy chain member 2; kinesin heavy chain member 2A; kinesin heavy chain member 2A L homeolog; Kinesin, heavy chain, 2; kinesin-2; Kinesin-like protein KIF2A; kinesin-related protein XKIF2; Kns2; M-kinesin; RCJMB04_9a7; UniProtKB:O00139}; Unknown (protein for MGC:133806); XELAEV_18008243mg
Rabbit
Affinity Chromatography
RUO
16563, 3796, 84391
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.59 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
O00139, P28740, Q9WV63
Kif2a
Recombinant fusion protein containing a sequence corresponding to amino acids 30-220 of human KIF2A (NP_0045112).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Promociones

Promociones

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado