Learn More
KIF1A Rabbit, anti-Human, Polyclonal, Proteintech
Código de producto. 18080546 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
18080546 150 μL 150 microlitros
18054715 20 μL 20 microlitros
2 options
Código de producto. 18080546
Marca: Proteintech 248411AP150UL
Rabbit Polyclonal Antibody
The KIF1A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KIF1A. This antibody recognizes human, mouse, rat antigen. The KIF1A antibody has been validated for the following applications: IF, WB, ELISA analysis.
Especificaciones
Antígeno KIF1A
Aplicaciones Western Blot, Immunofluorescence, ELISA
Concentración 300 μg/mL
Símbolos de los genes ATSV, C2orf20, HUNC 104, KIF1A, kinesin family member 1A, Kinesin like protein KIF1A, Microtubule based motor KIF1A, UNC104
Especie del huésped Rabbit
Cantidad 150 μL
Primario o secundario Primary
ID de gen (Entrez) 547
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C. Stable for one year after shipment.
Peso molecular del anticuerpo 150 kDa
Formulario Liquid
Isotype IgG