Learn More
Invitrogen™ KIF15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA557305
Descripción
Immunogen sequence: ESLLATEKVI SSLEKSRDSD KKVVADLMNQ IQELRTSVCE KTETIDTLKQ ELKDINCKYN SALVDREESR VLIKKQEVDI LD Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 70%.
Plus-end directed kinesin-like motor enzyme involved in mitotic spindle assembly. Target protein's mouse homolog associates with microtubulew and appears to play a role in mitosis in fibroblasts and in migration in developing neurons.
Especificaciones
| KIF15 | |
| Polyclonal | |
| Unconjugated | |
| KIF15 | |
| 3110023M17Rik; 3930402I10Rik; b2b1117.1aClo; b2b1117.1Clo; D330038N01; hKLP2; KIF15; kinesin 15; kinesin family member 15; kinesin-like 7; kinesin-like protein 2; Kinesin-like protein 7; kinesin-like protein KIF15; Klp2; Knsl7; N-10 kinesin; NY-BR-62; serologically defined breast cancer antigen NY-BR-62 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56992 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NS87 | |
| KIF15 | |
| Recombinant protein corresponding to Human KIF15. Recombinant protein control fragment (Product #RP-96412). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.