missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF14 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26000-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene encodes a member of the kinesin-3 superfamily of microtubule motor proteins. These proteins are involved in numerous processes including vesicle transport, chromosome segregation, mitotic spindle formation, and cytokinesis. In human HeLa-S3 and 293T cells, this protein is localized to the cytoplasm during interphase, to the spindle poles and spindle microtubules during mitosis, and to the midbody during cytokinesis. An internal motor domain displays microtubule-dependent ATPase activity, consistent with its function as a microtubule motor protein. Knockdown of this gene results in failed cytokinesis with endoreplication, which results in multinucleated cells. This gene has been identified as a likely oncogene in breast, lung and ovarian cancers, as well as retinoblastomas and gliomas. Alternative splicing results in multiple transcript variants.Especificaciones
| KIF14 | |
| Polyclonal | |
| Unconjugated | |
| KIF14 | |
| KIAA0042, KIF14, kinesin family member 14, Kinesin like protein KIF14 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9928 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q15058 | |
| KIF14 | |
| KIF14 Fusion Protein Ag23219 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido