missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ki67/MKI67 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NB500-170
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Ki67/MKI67 Polyclonal specifically detects Ki67/MKI67 in Human, Mouse, Rat, Porcine, Avian samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay.
Especificaciones
| Ki67/MKI67 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, antiKi67, Ki67, Ki-67, Ki67 ihc, Ki-67 ihc, Ki67 mouse, Ki-67 mouse, Ki67 western blot, Ki-67 western blot, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105 | |
| Rabbit | |
| 359 kDa | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience | |
| 4288 | |
| Human, Mouse, Rat, Pig, Avian | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay | |
| 1.0 mg/mL | |
| Western Blot 1-2 ug/ml, Flow Cytometry reported in scientific literature (PMID 34946963), Immunohistochemistry 1:50-1:300, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunoprecipitation reported by customer review, Immunohistochemistry-Paraffin 1:50-1:300, Immunohistochemistry-Frozen 1:50-1:300. Use reported in scientific literature (PMID 22493503), Immunoblotting reported in scientific literature (PMID 24248265), Knockout Validated | |
| P46013, Q91VE6 | |
| MKI67 | |
| The immunogen for this KI67/MKI67 Antibody was made using a synthetic peptide from the internal region of Human KI67/MKI67, between amino acids: 1218-1232 1582-1596, 2310-2324 [Uniprot: P46013]. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, mouse, rat (pubmed 22521325) and porcine. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido