missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: ADVPVVRGKP TLRTRGVPAP AITRGRGGVT ARPVGVVVPR GTPTPRGVLS TRGPVSRGRG LLTPRARGVP PTGYRPPPPP PTQETYGEYD YDDGYGTAYD EQSYDS Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 93%.
As a RNA-binding protein, KHDRBS3 plays a role in the regulation of alternative splicing and influences mRNA splice site selection and exon inclusion. KHDRBS3 may play a role as a negative regulator of cell growth. It inhibits cell proliferation and involved in splice site selection of vascular endothelial growth factor. It induces an increased concentration-dependent incorporation of exon in CD44 pre-mRNA by direct binding to purine-rich exonic enhancer. RNA-binding abilities are down-regulated by tyrosine kinase PTK6. It also involved in post-transcriptional regulation of HIV-1 gene expression.
Especificaciones
Especificaciones
| Antígeno | KHDRBS3 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 0.1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | Khdrbs3 |
| N.º de referencia del gen | O75525 |
| Alias de gen | Etle; etoile; etoile, Sam68-like protein SLM-2; KH domain containing, RNA binding, signal transduction associated 3; KH domain-containing, RNA-binding, signal transduction-associated protein 3; KH RNA binding domain containing, signal transduction associated 3; KHDRBS3; RNA-binding protein Etoile; RNA-binding protein T-Star; rSLM-2; SALP; Sam68-like mammalian protein 2; Sam68-like phosphotyrosine protein; Sam68-like phosphotyrosine protein, T-STAR; SLM2; SLM-2; TSTAR; T-STAR |
| Símbolos de los genes | Khdrbs3 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
