missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KCNQ2 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15897083
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15897083 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15897083 Proveedor Invitrogen™ N.º de proveedor PA568676

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 93%.

KCNQs are members of the voltage-dependent non-inactivating potassium channel family. Currently there are five known KNCQs (KCNQ1-5) found in the central nervous system and KNCQ2 and 3 have demonstrated their importance in M-current activation. Studies have shown that KCNQ2 and KCNQ3 form heteromultimers that, when formed, substantially increase the M-current. Inhibition of M-current controls neuron excitability throughout the nervous system as well as the responsiveness to synaptic inputs. Genetic mutations in these proteins have been linked to disorders such as benign familial neonatal convulsions (BFNC), deafness, neuropathic pain and epilepsy.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno KCNQ2
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 2% sucrose and 0.09% sodium azide
génica KCNQ2
N.º de referencia del gen Q9Z351
Alias de gen BFNC; EBN; EBN1; ENB1; HNSPC; KCNA11; KCNQ2; KQT like 2; KQT2; KQT-like 2; KV7.2; KVEBN1; mKQT2.3; mKQT2.4; neuroblastoma-specific potassium channel subunit alpha KvLQT2; Nmf134; potassium channel subunit alpha KvLQT2; potassium channel, voltage gated KQT-like subfamily Q, member 2; potassium channel, voltage-gated KQT-like subfamily Q, member 2; potassium voltage-gated channel KQT-like subfamily member 2.3; potassium voltage-gated channel KQT-like subfamily member 2.4; potassium voltage-gated channel subfamily KQT member 2; potassium voltage-gated channel subfamily Q member 2; potassium voltage-gated channel, KQT-like subfamily, member 2; potassium voltage-gated channel, subfamily Q, member 2; voltage-gated potassium channel subunit Kv7.2
Símbolos de los genes KCNQ2
Especie del huésped Rabbit
Inmunógeno synthetic peptide directed towards the middle region of human KCNQ2 (aa 182-231).
Método de purificación Affinity Chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 16536
Especies diana Mouse
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.