missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNMB4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA567237
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: CWLSPALQDLQ ATEANCTVLS VQQIGEVFEC TFTCGADCRG TSQYPCVQV Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.
MaxiK channels are large conductance, voltage and calcium-sensitive potassium channels which are fundamental to the control of smooth muscle tone and neuronal excitability. MaxiK channels can be formed by 2 subunits: the pore-forming alpha subunit and the modulatory beta subunit. The protein encoded by this gene is an auxiliary beta subunit which slows activation kinetics, leads to steeper calcium sensitivity, and shifts the voltage range of current activation to more negative potentials than does the beta 1 subunit.
Especificaciones
| KCNMB4 | |
| Polyclonal | |
| Unconjugated | |
| KCNMB4 | |
| 1700058G18Rik; 2900045G12Rik; big potassium channel beta subunit 4; BK channel beta subunit 4; BK channel subunit beta-4; BKbeta4; calcium activated potassium channel beta 4 subunit; calcium-activated potassium channel subunit beta-4; Calcium-activated potassium channel, subfamily M subunit beta-4; charybdotoxin receptor subunit beta-4; hbeta4; k(VCA)beta-4; KCMB4; KCNMB4; large conductance calcium-dependent potassium ion channel beta 4 subunit; maxi K channel subunit beta-4; MaxiK channel beta-subunit 4; potassium calcium-activated channel subfamily M regulatory beta subunit 4; potassium channel subfamily M regulatory beta subunit 4; potassium large conductance calcium-activated channel, subfamily M, beta member 4; slo-beta-4; Slowpoke beta 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27345 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86W47 | |
| KCNMB4 | |
| Recombinant Human KCNMB4. Recombinant protein control fragment (Product #RP-107871). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido