missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNG4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5104026
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Antibody detects endogenous levels of total KCNG4.
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily G. This member functions as a modulatory subunit. The gene has strong expression in brain. Multiple alternatively spliced variants have been found in normal and cancerous tissues.
Especificaciones
| KCNG4 | |
| Polyclonal | |
| Unconjugated | |
| KCNG4 | |
| 4921535I01Rik; AW049024; KCNG3; Kcng4; KV6.3; KV6.4; potassium channel, voltage gated modifier subfamily G, member 4; potassium voltage-gated channel modifier subfamily G member 4; potassium voltage-gated channel subfamily G member 4; potassium voltage-gated channel, subfamily G, member 4; voltage-gated potassium channel Kv6.3; voltage-gated potassium channel subunit Kv6.3; voltage-gated potassium channel subunit Kv6.4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 307900, 66733, 93107 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q80XM3, Q8TDN1 | |
| KCNG4 | |
| A synthesized peptide derived from human KCNG4(Accession Q8TDN1), corresponding to amino acid residues P294-I344. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido