missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCC2 Recombinant Rabbit Monoclonal Antibody (HL2625)
KCC2 Recombinant Rabbit Monoclonal Antibody (HL2625)
Marca: Invitrogen MA556771
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
KCC2 Recombinant Monoclonal Antibody for WB, IHC (P)
Especificaciones
| KCC2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q63633, Q91V14 | |
| SLC12A5 | |
| Synthetic peptide encompassing a sequence within the Intracellular domain of mouse KCC2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2625 | |
| Unconjugated | |
| SLC12A5 | |
| EIEE34; EIG14; Electroneutral potassium-chloride cotransporter 2; erythroid K-Cl cotransporter 2; furosemide-sensitive K-Cl cotransporter; hKCC2; KCC 2; Kcc2; KCC2a variant 1; KCC2a-S25 variant 1; KCC2b variant 2; K-Cl cotransporter 2; KIAA1176; mKCC2; mKIAA1176; neuronal K-Cl cotransporter; neuronal-specific K-Cl cotransporter; rKCC2; S12A5; SLC12A5; solute carrier family 12 (potassium/chloride transporter), member 5; solute carrier family 12 (potassium-chloride transporter), member 5; solute carrier family 12 member 5; solute carrier family 12, (potassium-chloride transporter) member 5; solute carrier family 12, member 5 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 171373, 57138 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido