Learn More
Invitrogen™ KCC2 Monoclonal Antibody (N1/12), PerCP
GREENER_CHOICE
Código de producto. 17848402
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
17848402 100 μg 100 microgramos
1 options
Código de producto. 17848402
Marca: Invitrogen™ MA545497
Mouse Monoclonal Antibody
1 μg/mL of MA5-45497 was sufficient for detection of KCC2 in 10 μg of rat brain lysate by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 140kDa. This antibody was formerly sold as clone S1-12.
Mediates electroneutral potassium-chloride cotransport in mature neurons. Transport occurs under isotonic conditions, but is activated 20-fold by cell swelling. Important for Cl(-) homeostasis in neurons.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno KCC2
Aplicaciones Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon N1/12
Concentración 1 mg/mL
Conjugado PerCP
Formulación 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
génica SLC12A5
N.º de referencia del gen Q63633, Q91V14, Q9H2X9
Alias de gen EIEE34; EIG14; Electroneutral potassium-chloride cotransporter 2; erythroid K-Cl cotransporter 2; furosemide-sensitive K-Cl cotransporter; hKCC2; KCC 2; Kcc2; KCC2a variant 1; KCC2a-S25 variant 1; KCC2b variant 2; K-Cl cotransporter 2; KIAA1176; mKCC2; mKIAA1176; neuronal K-Cl cotransporter; neuronal-specific K-Cl cotransporter; rKCC2; S12A5; SLC12A5; solute carrier family 12 (potassium/chloride transporter), member 5; solute carrier family 12 (potassium-chloride transporter), member 5; solute carrier family 12 member 5; solute carrier family 12, (potassium-chloride transporter) member 5; solute carrier family 12, member 5
Símbolos de los genes SLC12A5
Especie del huésped Mouse
Inmunógeno Fusion protein amino acids 932-1043 corresponding to rat KCC2.
Método de purificación Protein G
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 171373, 57138, 57468
Especies diana Human, Mouse, Rat
Contenido y almacenamiento 4°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2a