missing translation for 'onlineSavingsMsg'
Learn More

Kappa/Lambda Antibody Cocktail, Invitrogen™

Código de producto. 11325063
Change view
Click to view available options
Cantidad:
7 mL
Tamaño de la unidad:
7 ml
Código de producto. Cantidad unitSize
11325063 7 mL 7 ml
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11325063

Marca: Invitrogen MA514434

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Antibody Cocktail

This product is a cocktail containing mouse monoclonal antibody MA5-12117 (kappa) and rabbit polyclonal PA5-16651 (lambda). Kappa-lambda double staining contributes to the observation of clonality of the infiltrate. This product is provided at an optimal concentration in an easy-to-use format for demonstrating clonality. A ready-to-use MultiVision polymer detection system consisting of an anti-mouse (HRP) + anti-rabbit (AP) is available (Thermo Scientific Product No. TL-012-MHRA). This cocktail is specially formulated from MultiVision polymers that provide increased sensitivity, time-savings and detection simplicity. The MultiVision AP and HRP polymers are innovative, patented technology. The small amino acid polymer subunits minimize conflicts in binding the target protein resulting in more consistent staining and better signal amplification. Ultimately, this gives the user higher sensitivity and antibody efficiency as well as better signal-to-noise ratios. This system is biotin-free, which prevents background staining found with traditional biotin-based methods. MultiVision polymer detection system is a simple and robust one-step polymer detection system for double staining of kappa light chain and lambda light chain in red and blue, respectively. The MultiVision polymer detection system yields high quality double staining in less than 2 hours on formalin-fixed and paraffin-embedded tissue sections.

The antibodies to lambda and kappa light chain are reported as useful in the identification of leukemias, plasmacytomas and certain non-Hodgkin lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Kappa/Lambda
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Cocktail
Clon L1C1
Conjugado Unconjugated
Formulación PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
génica IGKC
N.º de referencia del gen P01834
Alias de gen HCAK1; IGKC; IGKCD; IGLV; IGLV@; immunoglobulin kappa constant; immunoglobulin lambda variable cluster; Km
Símbolos de los genes IGKC
Especie del huésped Mixed
Inmunógeno Kappa: B lymphoma cells. Lambda: light chains purified from human Bence-Jones proteins
Método de purificación purified
Cantidad 7 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3514
Especies diana Human
Contenido y almacenamiento 4° C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.