missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KAP3 Recombinant Rabbit Monoclonal Antibody (28E9)
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen™ MA550028
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
KAP3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
The small G protein GDP dissociation stimulator (smg GDS) is a regulator protein having two activities on a group of small G proteins including the Rho and Rap1 family members and Ki-Ras; one is to stimulate their GDP/GTP exchange reactions, and the other is to inhibit their interactions with membranes. The protein encoded by this gene contains 9 'Armadillo' repeats and interacts with the smg GDS protein through these repeats. This protein, which is highly concentrated around the endoplasmic reticulum, is phosphorylated by v-src, and this phosphorylation reduces the affinity of the protein for smg GDS. It is thought that this protein serves as a linker between human chromosome-associated polypeptide (HCAP) and KIF3A/B, a kinesin superfamily protein in the nucleus, and that it plays a role in the interaction of chromosomes with an ATPase motor protein. Several transcript variants encoding different isoforms have been found for this gene.
Especificaciones
| KAP3 | |
| Recombinant Monoclonal | |
| 0.61 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q92845 | |
| KIFAP3 | |
| A synthesized peptide derived from human KIFAP3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 28E9 | |
| Unconjugated | |
| KIFAP3 | |
| dJ190I16.1; FLA3; KAP-1; KAP3; KAP-3; KIF3AP; Kifap3; kinesin associated protein 3; kinesin-associated protein 3; small G protein GDP dissociation stimulator; SMAP; Smg GDS-associated protein; Smg-GDS | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22920 | |
| -20°C or -80°C if preferred | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido