missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KALRN Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Marca: Novus Biologicals NB100-41371
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
KALRN Polyclonal antibody specifically detects KALRN in Human, Mammalia samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA
Especificaciones
| KALRN | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA | |
| ARHGEF24, DUETFLJ16443, DUO, EC 2.7.11.1, FLJ12332, FLJ18623, HAPIP, huntingtin-associated protein interacting protein (duo), Huntingtin-associated protein-interacting protein, Kalirin, kalirin, RhoGEF kinase, Protein Duo, serine/threonine kinase with Dbl- and pleckstrin homology domains, Serine/threonine-protein kinase with Dbl- and pleckstrin homology domain, TRADFLJ18196 | |
| Peptide with sequence C-RWHLGPGDPFSTYV, from the C Terminus of the protein sequence according to NP_003938.1; NP_001309918.1; NP_001309919.1; NP_001309920.1; NP_001309921.1. | |
| 0.1 mg | |
| Neuroscience | |
| 8997 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA | |
| 0.5 mg/mL | |
| Flow Cytometry 10 μg/mL, Immunohistochemistry 4 - 5 μg/mL, Immunohistochemistry-Paraffin 4 - 5 μg/mL, Peptide ELISA Detection Limit 1:16000 | |
| O60229 | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mammalia | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido