missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ K-cadherin Recombinant Rabbit Monoclonal Antibody (23GB5060), Invitrogen™

Descripción
Cadherins constitute a family of transmembrane glycoproteins involved in Ca2+-dependent cell-cell interactions. The members of this family are differentially expressed in various tissues. They function in the maintenance of tissue integrity and morphogenesis. Cadherins are divided into type I and type II subgroups. Type I cadherins include epithelial cadherin (E-cadherin, cadherin-1 or uvomorulin), neural cadherin (N-cadherin or cadherin-2), placental cadherin (P-cadherin or cadherin-3) and retinal cadherin (R-cadherin or cadherin-4), whereas kidney cadherin (K-cadherin or cadherin-6) and osteoblast cadherin (OB-cadherin or cadherin-11) are type II cadherins. Cadherin-6 is expressed in the proximal tubule epithelium of the kidney from which most renal cell carcinomas are derived.
Especificaciones
Especificaciones
| Antígeno | K-cadherin |
| Aplicaciones | Flow Cytometry, Western Blot, Immunocytochemistry |
| Clasificación | Recombinant Monoclonal |
| Clon | 23GB5060 |
| Concentración | 0.2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | CDH6 |
| N.º de referencia del gen | P55285, P97326 |
| Alias de gen | CAD6; CADH6; cadherin 6; cadherin 6, type 2, K-cadherin; cadherin 6, type 2, K-cadherin (fetal kidney); Cadherin6; cadherin-6; Cdh6; k cadherin; KCAD; K-cadherin; Kidney cadherin |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?