missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JPH2 Monoclonal Antibody (OTI2E5), TrueMAB™, OriGene
Descripción
Junctional complexes between the plasma membrane (PM) and endoplasmic/sarcoplasmic reticulum (ER/SR) are a common feature of all excitable cell types and mediate cross talk between cell surface and intracellular ion channels. Junctophilins (JPs) are important components of the junctional complexes. JPs are composed of a carboxy-terminal hydrophobic segment spanning the ER/SR membrane and a remaining cytoplasmic domain that shows specific affinity for the PM. Four JPs have been identified as tissue-specific subtypes derived from different genes: JPH1 is expressed in skeletal muscle, JPH2 is detected throughout all muscle cell types, and JPH3 and JPH4 are predomitly expressed in the brain and contribute to the subsurface cistern formation in neurons. JPH2-null mice died of embryonic cardiac arrest and human patients with mutations in the JPH2 gene showed hypertrophic cardiomyopathy, demonstrating the importance of this protein. Multiple isoforms of JPH2 are known to exist.
Especificaciones
Especificaciones
| Antígeno | JPH2 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI2E5 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | JPH2 |
| N.º de referencia del gen | Q9BR39 |
| Alias de gen | CMH17, JP-2, JP2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?